missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthoyl Cyanide 98.0+%, TCI America™
$322.48 - $322.48
Chemical Identifiers
| CAS | 14271-86-4 |
|---|---|
| Molecular Formula | C12H7NO |
| Molecular Weight (g/mol) | 181.19 |
| MDL Number | MFCD00060239 |
| InChI Key | RITADAJGEQPFIR-UHFFFAOYSA-N |
| Synonym | 1-Naphthalenecarbonyl Cyanide |
| PubChem CID | 602423 |
| IUPAC Name | naphthalene-1-carbonyl cyanide |
| SMILES | O=C(C#N)C1=CC=CC2=CC=CC=C12 |
Chemical Identifiers
| 14271-86-4 | |
| 181.19 | |
| RITADAJGEQPFIR-UHFFFAOYSA-N | |
| 602423 | |
| O=C(C#N)C1=CC=CC2=CC=CC=C12 |
| C12H7NO | |
| MFCD00060239 | |
| 1-Naphthalenecarbonyl Cyanide | |
| naphthalene-1-carbonyl cyanide |
Specifications
| 14271-86-4 | |
| Yellow | |
| C12H7NO | |
| 5 g | |
| 1-Naphthalenecarbonyl Cyanide | |
| O=C(C#N)C1=CC=CC2=CC=CC=C12 | |
| 181.19 | |
| 181.19 | |
| Crystalline Powder |
| 100°C | |
| 182°C | |
| MFCD00060239 | |
| 3439 | |
| RITADAJGEQPFIR-UHFFFAOYSA-N | |
| naphthalene-1-carbonyl cyanide | |
| 602423 | |
| ≥98.0% (GC,N) | |
| 1-Naphthoyl Cyanide |
Safety and Handling
TSCA : No