missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoyl Cyanide 98.0+%, TCI America™

Catalog No. N00495G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N00495G Supplier TCI America Supplier No. N00495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14271-86-4
Molecular Formula C12H7NO
Molecular Weight (g/mol) 181.19
MDL Number MFCD00060239
InChI Key RITADAJGEQPFIR-UHFFFAOYSA-N
Synonym 1-Naphthalenecarbonyl Cyanide
PubChem CID 602423
IUPAC Name naphthalene-1-carbonyl cyanide
SMILES O=C(C#N)C1=CC=CC2=CC=CC=C12

Specifications

Melting Point 100°C
Color Yellow
Boiling Point 182°C
Quantity 5 g
UN Number 3439
Formula Weight 181.19
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthoyl Cyanide