Learn More
1-Naphthoyl chloride, 98%
CAS: 879-18-5 | C11H7ClO | 190.63 g/mol
Supplier: Thermo Scientific Chemicals A1446009
Description
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, amines, alcohols, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
| 1-Naphthoyl chloride | |
| 16°C to 19°C | |
| 118°C (0.1 mmHg) | |
| Pungent | |
| 1.6525 | |
| 10 g | |
| 775785 | |
| 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride | |
| NSNPSJGHTQIXDO-UHFFFAOYSA-N | |
| naphthalene-1-carbonyl chloride | |
| 70146 | |
| 98% |
| 879-18-5 | |
| 1.265 | |
| >110°C (230°F) | |
| C11H7ClO | |
| MFCD00004002 | |
| UN3265 | |
| Moisture Sensitive | |
| Reacts with water. | |
| ClC(=O)C1=C2C=CC=CC2=CC=C1 | |
| 190.63 | |
| 190.63 |
Chemical Identifiers
| 879-18-5 | |
| 190.63 | |
| NSNPSJGHTQIXDO-UHFFFAOYSA-N | |
| 70146 | |
| ClC(=O)C1=C2C=CC=CC2=CC=C1 |
| C11H7ClO | |
| MFCD00004002 | |
| 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride | |
| naphthalene-1-carbonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-903-9
TSCA : Yes
RUO – Research Use Only