missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoyl chloride, 98%

Catalog No. AAA1446030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1446030 250 g
AAA1446009 10 g
AAA1446018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1446030 Supplier Thermo Scientific Chemicals Supplier No. A1446030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 879-18-5 | C11H7ClO | 190.63 g/mol

1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, amines, alcohols, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 879-18-5
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.63
MDL Number MFCD00004002
InChI Key NSNPSJGHTQIXDO-UHFFFAOYSA-N
Synonym 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride
PubChem CID 70146
IUPAC Name naphthalene-1-carbonyl chloride
SMILES ClC(=O)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 16°C to 19°C
Density 1.265
Boiling Point 118°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6525
Quantity 250 g
UN Number UN3265
Beilstein 775785
Sensitivity Moisture Sensitive
Solubility Information Reacts with water.
Formula Weight 190.63
Percent Purity 98%
Chemical Name or Material 1-Naphthoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-903-9
TSCA Yes

RUO – Research Use Only