Learn More
1-Naphthalenemethylamine, 98+%
CAS: 118-31-0 | C11H12N | 158.22 g/mol
Supplier: Thermo Scientific Chemicals L0354106
Description
1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.
Solubility
Miscible with ethanol, ether and carbon disulfide.
Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Specifications
| 1-Naphthalenemethylamine | |
| 1.075 | |
| >110°C (230°F) | |
| 1.643 | |
| 5 g | |
| 2206459 | |
| 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride | |
| NVSYANRBXPURRQ-UHFFFAOYSA-O | |
| naphthalen-1-ylmethanamine | |
| 8355 | |
| ≥98% |
| 118-31-0 | |
| 289°C to 292°C | |
| C11H12N | |
| MFCD00004048 | |
| UN2735 | |
| Air sensitive; May form a precipitate | |
| Miscible with ethanol,ether and carbon disulfide. | |
| [NH3+]CC1=C2C=CC=CC2=CC=C1 | |
| 158.22 | |
| 157.22 |
Chemical Identifiers
| 118-31-0 | |
| 158.22 | |
| NVSYANRBXPURRQ-UHFFFAOYSA-O | |
| 8355 | |
| [NH3+]CC1=C2C=CC=CC2=CC=C1 |
| C11H12N | |
| MFCD00004048 | |
| 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride | |
| naphthalen-1-ylmethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 204-244-0
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only