missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethylamine, 98+%

Catalog No. AAL0354103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0354103 1 g
AAL0354106 5 g
AAL0354114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0354103 Supplier Thermo Scientific Chemicals Supplier No. L0354103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-31-0 | C11H12N | 158.22 g/mol

1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.

Solubility
Miscible with ethanol, ether and carbon disulfide.

Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.

Chemical Identifiers

CAS 118-31-0
Molecular Formula C11H12N
Molecular Weight (g/mol) 158.22
MDL Number MFCD00004048
InChI Key NVSYANRBXPURRQ-UHFFFAOYSA-O
Synonym 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
PubChem CID 8355
IUPAC Name naphthalen-1-ylmethanamine
SMILES [NH3+]CC1=C2C=CC=CC2=CC=C1

Specifications

Density 1.075
Boiling Point 289°C to 292°C
Flash Point >110°C (230°F)
Refractive Index 1.643
Quantity 1 g
UN Number UN2735
Beilstein 2206459
Sensitivity Air sensitive; May form a precipitate
Solubility Information Miscible with ethanol,ether and carbon disulfide.
Formula Weight 157.22
Percent Purity ≥98%
Chemical Name or Material 1-Naphthalenemethylamine
Show More Show Less
Hazard Category H315-H318
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 204-244-0
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only