missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethylamine, 97%

Catalog No. AC155580010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155580010 1 g Glass bottle
AC155580050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155580010 Supplier Thermo Scientific Chemicals Supplier No. 155580010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-31-0 | C11H12N | 158.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-31-0
Molecular Formula C11H12N
Molecular Weight (g/mol) 158.22
MDL Number MFCD00004048
InChI Key NVSYANRBXPURRQ-UHFFFAOYSA-O
Synonym 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
PubChem CID 8355
IUPAC Name naphthalen-1-ylmethanamine
SMILES [NH3+]CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.0730g/mL
Boiling Point 290°C to 293°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.643
Linear Formula C10H7CH2NH2
Quantity 1 g
Beilstein 12, 1316
Specific Gravity 1.073
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol,ether,carbon disulfide,sulfu,ric acid
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Naphthalenemethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
EINECSNumber 204-244-

RUO – Research Use Only