missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. N06301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06301G Supplier TCI America Supplier No. N06301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13922-41-3
Molecular Formula C10H9BO2
Molecular Weight (g/mol) 171.99
MDL Number MFCD00019722
InChI Key HUMMCEUVDBVXTQ-UHFFFAOYSA-N
Synonym 1-naphthaleneboronic acid, 1-naphthylboronic acid, naphthalene-1-boronic acid, naphthalenyl-1-boronic acid, 1-naphthalene boronic acid, 1-naphthaleneboronicacid, 1-naphthyleneboronic acid, boronic acid, naphthalenyl, 1-borononaphthalene, naphthylboronic acid
PubChem CID 254532
IUPAC Name (naphthalen-1-yl)boronic acid
SMILES OB(O)C1=CC=CC2=CC=CC=C12

Specifications

Melting Point 211°C
Quantity 1 g
Formula Weight 171.99
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthaleneboronic Acid (contains varying amounts of Anhydride)