missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneboronic acid, 97%

CAS: 13922-41-3 | C10H9BO2 | 171.99 g/mol

Supplier:  Thermo Scientific Chemicals B2121906

 View more versions of this product

Catalog No. AAB2121906


Only null left
Explore available promotions

Description

Description

1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.
Specifications

Specifications

1-Naphthaleneboronic acid
204°C to 210°C
MFCD00019722
2937504
Soluble in methanol. Insoluble in water.
OB(O)C1=CC=CC2=CC=CC=C12
171.99
171.99
13922-41-3
C10H9BO2
5 g
1-naphthaleneboronic acid, 1-naphthylboronic acid, naphthalene-1-boronic acid, naphthalenyl-1-boronic acid, 1-naphthalene boronic acid, 1-naphthaleneboronicacid, 1-naphthyleneboronic acid, boronic acid, naphthalenyl, 1-borononaphthalene, naphthylboronic acid
HUMMCEUVDBVXTQ-UHFFFAOYSA-N
naphthalen-1-ylboronic acid
254532
97%

Chemical Identifiers

13922-41-3
171.99
HUMMCEUVDBVXTQ-UHFFFAOYSA-N
254532
OB(O)C1=CC=CC2=CC=CC=C12
C10H9BO2
MFCD00019722
1-naphthaleneboronic acid, 1-naphthylboronic acid, naphthalene-1-boronic acid, naphthalenyl-1-boronic acid, 1-naphthalene boronic acid, 1-naphthaleneboronicacid, 1-naphthyleneboronic acid, boronic acid, naphthalenyl, 1-borononaphthalene, naphthylboronic acid
naphthalen-1-ylboronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only