missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneboronic acid, 97%

Catalog No. AAB2121914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2121914 25 g
AAB2121903 1 g
AAB2121906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2121914 Supplier Thermo Scientific Chemicals Supplier No. B2121914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13922-41-3 | C10H9BO2 | 171.99 g/mol

1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.

Chemical Identifiers

CAS 13922-41-3
Molecular Formula C10H9BO2
Molecular Weight (g/mol) 171.99
MDL Number MFCD00019722
InChI Key HUMMCEUVDBVXTQ-UHFFFAOYSA-N
Synonym 1-naphthaleneboronic acid, 1-naphthylboronic acid, naphthalene-1-boronic acid, naphthalenyl-1-boronic acid, 1-naphthalene boronic acid, 1-naphthaleneboronicacid, 1-naphthyleneboronic acid, boronic acid, naphthalenyl, 1-borononaphthalene, naphthylboronic acid
PubChem CID 254532
IUPAC Name naphthalen-1-ylboronic acid
SMILES OB(O)C1=CC=CC2=CC=CC=C12

Specifications

Melting Point 204°C to 210°C
Quantity 25 g
Beilstein 2937504
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 171.99
Percent Purity 97%
Chemical Name or Material 1-Naphthaleneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only