missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyltheobromine, 98+%

Catalog No. AAL13597MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13597MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL13597MD Supplier Thermo Scientific Chemicals Supplier No. L13597MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1028-33-7 | C13H20N4O2 | 264.33 g/mol

A vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 1028-33-7
Melting Point 80°C to 81°C
Molecular Formula C13H20N4O2
Quantity 250 mg
MDL Number MFCD00041424
Beilstein 270632
Merck Index 14,7127
Synonym pentifylline, 1-hexyltheobromine, 1-n-hexyltheobromine, hexyltheobromine, cosaldon, pentifyllin, cosadon, theobromine, 1-hexyl, sk 7 pharmaceutical, 1-hexyl-3,7-dimethylxanthine
InChI Key MRWQRJMESRRJJB-UHFFFAOYSA-N
SMILES CCCCCCN1C(=O)N(C)C2=C(N(C)C=N2)C1=O
IUPAC Name 1-hexyl-3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Weight (g/mol) 264.33
PubChem CID 70569
Formula Weight 264.33
Percent Purity ≥98%
Chemical Name or Material 1-n-Hexyltheobromine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-842-0
RTECSNumber UO8450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only