missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium iodide, 98%

Catalog No. AAH5948706
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5948706 5 g
AAH5948718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5948706 Supplier Thermo Scientific Chemicals Supplier No. H5948706

CAS: 178631-05-5 | C10H19IN2 | 294.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 178631-05-5
Molecular Formula C10H19IN2
Molecular Weight (g/mol) 294.18
MDL Number MFCD12964976
InChI Key CZIUVCSYOGFUPH-UHFFFAOYSA-M
Synonym 1-hexyl-3-methylimidazolium iodide, 1-hexyl-3-methyl-1h-imidazol-3-ium iodide, 1-n-hexyl-3-methylimidazolium iodide, dsstox_cid_27927, dsstox_rid_82680, dsstox_gsid_47951, 1-methyl-3-hexylimidazolium iodide, 1-hexyl-3-me-thylimidazolium iodide, 1h-imidazolium,1-hexyl-3-methyl-,iodide, 1-hexyl-3-methylimidazolium iodide hplc
PubChem CID 12036651
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;iodide
SMILES CCCCCCN1C=C[N+](=C1)C.[I-]

Specifications

Color Colorless to Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 294.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only