missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%

Catalog No. AAH2727206
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2727206 5 g
AAH2727218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2727206 Supplier Thermo Scientific Chemicals Supplier No. H2727206

CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 304680-35-1
Molecular Formula C10H19F6N2P
Molecular Weight (g/mol) 312.24
MDL Number MFCD03093296
InChI Key YPWSSSRXUOQNMQ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
PubChem CID 2734175
SMILES F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1

Specifications

Refractive Index 1.419
Quantity 5 g
IUPAC Name 1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Formula Weight 312.24
Percent Purity 99%
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only