missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butylpyridinium chloride, 98%

Catalog No. p-7049727
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1974004 2 g
AA1974009 10 g
AA1974018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1974004 Supplier Thermo Scientific Chemicals Supplier No. 01974004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1124-64-7 | C9H14ClN | 171.668 g/mol

The derivatives of 1-n-Butylpyridinium chloride is used in spent fuel reprocessing, particularly in the extraction of high-level radioactive aqueous waste from the processing of nuclear fuel. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1124-64-7
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.668
MDL Number MFCD00060047
InChI Key POKOASTYJWUQJG-UHFFFAOYSA-M
Synonym 1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
PubChem CID 2734171
IUPAC Name 1-butylpyridin-1-ium;chloride
SMILES CCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Melting Point 131°C to 132°C
Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 171.67
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-n-Butylpyridinium chloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only