missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butylcyclohexanol, 98%

Catalog No. p-7025205
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2057906 5 g
AAB2057914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2057906 Supplier Thermo Scientific Chemicals Supplier No. B2057906
Only null left

CAS: 5445-30-7 | C10H20O | 156.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5445-30-7
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00021404
InChI Key RCHLXMOXBJRGNX-UHFFFAOYSA-N
Synonym 1-butylcyclohexanol, cyclohexanol, 1-butyl, 1-n-butylcyclohexanol, butylcyclohexanol, 1-butyl-cyclohexanol, 1-butyl-cyclohexan-1-ol
PubChem CID 138505
IUPAC Name 1-butylcyclohexan-1-ol
SMILES CCCCC1(CCCCC1)O

Specifications

Quantity 5 g
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 1-n-Butylcyclohexanol
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only