missing translation for 'onlineSavingsMsg'
Learn More

1-(n-Butyl)imidazole, 99%

Catalog No. p-7044688
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0779314 25 g
AAL0779322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0779314 Supplier Thermo Scientific Chemicals Supplier No. L0779314
Only null left

CAS: 4316-42-1 | C7H12N2 | 124.19 g/mol

1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.

Solubility
Soluble in Chloroform, Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 4316-42-1
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00042753
InChI Key MCMFEZDRQOJKMN-UHFFFAOYSA-N
Synonym 1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
PubChem CID 61347
IUPAC Name 1-butyl-1H-imidazole
SMILES CCCCN1C=CN=C1

Specifications

Density 0.948
Boiling Point 257°C
Flash Point 145°C (293°F)
Odor Amine-like
Refractive Index 1.48
Quantity 25 g
UN Number UN2690
Beilstein 109499
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 124.19
Percent Purity 99%
Chemical Name or Material 1-(n-Butyl)imidazole
Show More Show Less
Hazard Category H315-H318-H330-H335
Hazard Statement GHS H Statement
H330-H318-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-N-BUTYL IMIDAZOLE
EINECSNumber 224-335-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only