missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-4-methylpyridinium tetrafluoroborate, 99%

Catalog No. AAH2710006
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2710006 5 g
AAH2710018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2710006 Supplier Thermo Scientific Chemicals Supplier No. H2710006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 343952-33-0 | C10H16BF4N | 237.049 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 343952-33-0
Molecular Formula C10H16BF4N
Molecular Weight (g/mol) 237.049
MDL Number MFCD03095460
InChI Key VISYYHYJMCAKAF-UHFFFAOYSA-N
Synonym 1-butyl-4-methylpyridinium tetrafluoroborate, 1-butyl-4-methylpyridin-1-ium tetrafluoroborate, acmc-1adzo, dsstox_cid_29131, dsstox_rid_83350, dsstox_gsid_49275, 1-butyl-4-picolinium tetrafluoroborate, pyridinium, 1-butyl-4-methyl-, tetrafluoroborate 1-, 1-butyl-4-methylpyridinium tetrafluoroborate t
PubChem CID 11492406
IUPAC Name 1-butyl-4-methylpyridin-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCC[N+]1=CC=C(C=C1)C

Specifications

Density 1.2
Refractive Index 1.4
Quantity 5 g
Formula Weight 237.06
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-4-methylpyridinium tetrafluoroborate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only