missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium methanesulfonate, 99%

Catalog No. AAH2733618
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2733618 50 g
AAH2733606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2733618 Supplier Thermo Scientific Chemicals Supplier No. H2733618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 342789-81-5 | C9H18N2O3S | 234.314 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 342789-81-5
Molecular Formula C9H18N2O3S
Molecular Weight (g/mol) 234.314
MDL Number MFCD06798173
InChI Key PUHVBRXUKOGSBC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methanesulfonate, 1-butyl-3-methylimidazolium mesylate, acmc-20aowi, c4mim mesylate, basionics™ st 78, dsstox_cid_27888, dsstox_rid_82641, dsstox_gsid_47912, 1-butyl-3-methylimidazolium methylsulfonate, 1-n-butyl-3-methylimidazolium methanesulfonate
PubChem CID 11492381
IUPAC Name 1-butyl-3-methylimidazol-3-ium;methanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]

Specifications

Melting Point 75°C to 80°C
Flash Point 119°C (246°F)
Quantity 50 g
UN Number UN3261
Sensitivity Hygroscopic
Formula Weight 234.32
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium methanesulfonate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only