missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%

Catalog No. aal1908606
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAL1908606 5 g
AAL1908614 25 g
AAL1908622 100 g
3 options

Catalog No. AAL1908606

Supplier: Thermo Scientific Chemicals L1908606

Only null left

CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol

1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.

Solubility
Miscible with dichloromethane, chloroform, ethyl acetate. Immiscible with water, diethyl ether and hexane.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 174501-64-5
Molecular Formula C8H15F6N2P
Molecular Weight (g/mol) 284.19
MDL Number MFCD03093295
InChI Key IXQYBUDWDLYNMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
PubChem CID 2734174
IUPAC Name 1-butyl-3-methylimidazol-3-ium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Melting Point 8°C to 10°C
Density 1.365
Boiling Point >350°C
Refractive Index 1.4115
Quantity 5 g
Sensitivity Hygroscopic
Merck Index 14,1581
Solubility Information Miscible with dichloromethane,chloroform,ethyl acetate. Immiscible with water,diethyl ether and hexane.
Formula Weight 284.19
Percent Purity ≥98%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium hexafluorophosphate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only