missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium dicyanamide, 97%

CAS: 448245-52-1 | C10H15N5 | 205.265 g/mol

$225.96 - $1419.30

Chemical Identifiers

CAS 448245-52-1
Molecular Formula C10H15N5
Molecular Weight (g/mol) 205.265
MDL Number MFCD07784457
InChI Key ICIVTHOGIQHZRY-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
PubChem CID 11171745
IUPAC Name 1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide
SMILES CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5917506
View Documents
Thermo Scientific Chemicals
H5917506
5 g
Each for $225.96
Only null left
 
AAH5917518
View Documents
Thermo Scientific Chemicals
H5917518
50 g
Each for $1,419.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

448245-52-1
205.265
ICIVTHOGIQHZRY-UHFFFAOYSA-N
11171745
CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N
C10H15N5
MFCD07784457
1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide

Specifications

448245-52-1
C10H15N5
5 g
1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N
205.265
205.26
Liquid
Yellow
MFCD07784457
Moisture sensitive
ICIVTHOGIQHZRY-UHFFFAOYSA-N
1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide
11171745
97%
1-n-Butyl-3-methylimidazolium dicyanamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only