missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium chloride, 96%

Catalog No. p-7047785
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1974909 10 g
AAL1974918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1974909 Supplier Thermo Scientific Chemicals Supplier No. L1974909
Only null left

CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol

1-n-Butyl-3-methylimidazolium chloride is used as a solvent for organic reactions. It is used in anion exchange reactions. It acts as a catalyst, electrolyte for batteries and metal recovery. It is also used in chromatographic separations. Further, it is used as an ionic liquids for catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium chloride is used as a solvent for organic reactions. It is used in anion exchange reactions. It acts as a catalyst, electrolyte for batteries and metal recovery. It is also used in chromatographic separations. Further, it is used as an ionic liquids for catalysis.

Solubility
Soluble in acetone, acetonitrile, hot ethyl acetate, isopropyl alcohol, methylene chloride and methanol. Insoluble in water, hexane and toluene,

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 79917-90-1
Molecular Formula C8H15ClN2
Molecular Weight (g/mol) 174.67
MDL Number MFCD03095425
InChI Key FHDQNOXQSTVAIC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461
PubChem CID 2734161
SMILES [Cl-].CCCCN1C=C[N+](C)=C1

Specifications

Melting Point ∼73°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,acetonitrile,hot ethyl acetate,isopropyl alcohol,methylene chloride and methanol. Insoluble in water,hexane and toluene,
IUPAC Name 1-butyl-3-methylimidazol-3-ium;chloride
Formula Weight 174.67
Percent Purity 96%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium chloride
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only