missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium bromide, 99%

Catalog No. p-7029708
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2720106 5 g
AAH2720118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2720106 Supplier Thermo Scientific Chemicals Supplier No. H2720106
Only null left

CAS: 85100-77-2 | C8H15BrN2 | 219.126 g/mol

It is an ionic liquid used in a Heck reaction. It is also used in assessing the factors responsible for ionic liquid toxicity to aquatic organisms via quantitative structure-property relationship modeling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an ionic liquid used in a Heck reaction. It is also used in assessing the factors responsible for ionic liquid toxicity to aquatic organisms via quantitative structure-property relationship modeling

Solubility
Miscible in water, methanol and dichloromethane. Immiscible in acetone, toluene, ethyl acetate and diethyl ether.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 85100-77-2
Molecular Formula C8H15BrN2
Molecular Weight (g/mol) 219.126
MDL Number MFCD03427611
InChI Key KYCQOKLOSUBEJK-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium bromide, 3-butyl-1-methyl-1h-imidazol-3-ium bromide, bmimbr, 1-n-butyl-3-methylimidazolium bromide, 1-butyl-3-methyl-1h-imidazol-3-ium bromide, bmim br, c4mim bromide, ksc491c7d, 1-butyl-3-methylimidazoliumbromide
PubChem CID 2734236
IUPAC Name 1-butyl-3-methylimidazol-3-ium;bromide
SMILES CCCCN1C=C[N+](=C1)C.[Br-]

Specifications

Melting Point 65°C to 75°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Miscible in water,methanol and dichloromethane. Immiscible in acetone,toluene,ethyl acetate and diethyl ether.
Formula Weight 219.12
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium bromide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only