missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-2,3-dimethylimidazolium tetrafluoroborate, 99%

CAS: 402846-78-0 | C9H17BF4N2 | 240.05 g/mol

$104.67 - $390.14

Chemical Identifiers

CAS 402846-78-0
Molecular Formula C9H17BF4N2
Molecular Weight (g/mol) 240.05
MDL Number MFCD03427618
InChI Key VCAIYEJBOWHUGP-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b
PubChem CID 2734244
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2714106
View Documents
Thermo Scientific Chemicals
H2714106
5 g
Each for $104.67
Only null left
 
AAH2714118
View Documents
Thermo Scientific Chemicals
H2714118
50 g
Each for $390.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

402846-78-0
240.05
VCAIYEJBOWHUGP-UHFFFAOYSA-N
2734244
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C
C9H17BF4N2
MFCD03427618
1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b
1-butyl-2,3-dimethylimidazol-3-ium;tetrafluoroborate

Specifications

402846-78-0
C9H17BF4N2
5 g
VCAIYEJBOWHUGP-UHFFFAOYSA-N
1-butyl-2,3-dimethylimidazol-3-ium;tetrafluoroborate
2734244
99%
1.198
MFCD03427618
1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C
240.05
240.05
1-n-Butyl-2,3-dimethylimidazolium tetrafluoroborate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only