missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-2,3-dimethylimidazolium tetrafluoroborate, 99%

Catalog No. AAH2714118
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2714118 50 g
AAH2714106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2714118 Supplier Thermo Scientific Chemicals Supplier No. H2714118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 402846-78-0 | C9H17BF4N2 | 240.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402846-78-0
Molecular Formula C9H17BF4N2
Molecular Weight (g/mol) 240.05
MDL Number MFCD03427618
InChI Key VCAIYEJBOWHUGP-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b
PubChem CID 2734244
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Density 1.198
Quantity 50 g
Formula Weight 240.05
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-2,3-dimethylimidazolium tetrafluoroborate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only