missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, 98%

Catalog No. p-7029685
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2717706 5 g
AAH2717718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2717706 Supplier Thermo Scientific Chemicals Supplier No. H2717706

CAS: 223437-11-4 | C11H20F6N2O4S2 | 422.40 g/mol

1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.

Solubility
Miscible with methanol, acetonitrile, acetone, dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 223437-11-4
Molecular Formula C11H20F6N2O4S2
Molecular Weight (g/mol) 422.40
MDL Number MFCD07784447
InChI Key HSLXOARVFIWOQF-UHFFFAOYSA-N
Synonym 1-butyl-1-methylpyrrolidinium bis trifluoromethylsulfonyl imide, unii-fd74dkk1wh, fd74dkk1wh, 1-butyl-1-methylpyrrolidinium bis trifluoromethanesulfonyl imide, 1-butyl-1-methylpyrrolidin-1-ium bis trifluoromethyl sulfonyl amide, 1-butyl-1-methylpyrrolidin-1-ium; bistriflylimide anion, dsstox_cid_27944, dsstox_rid_82696, dsstox_gsid_47968, pyrrolidinium, 1-butyl-1-methyl-, salt with 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide 1:1
PubChem CID 11048104
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-butyl-1-methylpyrrolidin-1-ium
SMILES CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Solubility Information Miscible with methanol,acetonitrile,acetone,dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.
Formula Weight 422.42
Percent Purity 98%
Chemical Name or Material 1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only