missing translation for 'onlineSavingsMsg'
Learn More

1-Morpholino-1-cyclohexene 97.0+%, TCI America™

Catalog No. C095625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C095625ML 25 mL
C0956500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C095625ML Supplier TCI America Supplier No. C095625ML

Chemical Identifiers

CAS 670-80-4
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00006163
InChI Key IIQFBBQJYPGOHJ-UHFFFAOYSA-N
Synonym 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene
PubChem CID 69589
IUPAC Name 4-(cyclohexen-1-yl)morpholine
SMILES C1CCC(=CC1)N2CCOCC2

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 mL
Formula Weight 167.25
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Morpholino-1-cyclohexene
TSCA Yes
Recommended Storage Refrigerator