missing translation for 'onlineSavingsMsg'
Learn More

1-Methylurea 98.0+%, TCI America™

Catalog No. M045525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045525G 25 g
M0455500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M045525G Supplier TCI America Supplier No. M045525G

Chemical Identifiers

CAS 598-50-5
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
MDL Number MFCD00007950
InChI Key XGEGHDBEHXKFPX-UHFFFAOYSA-N
Synonym 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
PubChem CID 11719
ChEBI CHEBI:44383
IUPAC Name methylurea
SMILES CNC(=O)N

Specifications

Melting Point 99°C
Color White
Quantity 25 g
Formula Weight 74.08
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylurea
EINECSNumber (2)-3254
RTECSNumber YT7175000
TSCA Yes