missing translation for 'onlineSavingsMsg'
Learn More

1-(Methylsulfonyl)piperazine 98.0+%, TCI America™

Catalog No. M24375G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24375G Supplier TCI America Supplier No. M24375G

Chemical Identifiers

CAS 55276-43-2
Molecular Formula C5H13N2O2S
Molecular Weight (g/mol) 165.23
MDL Number MFCD02153225
InChI Key ZZAKLGGGMWORRT-UHFFFAOYSA-O
Synonym 1-methylsulfonyl piperazine, 1-methanesulfonylpiperazine, 1-methanesulfonyl-piperazine, 1-methylsulfonyl-piperazine, 1-methylsulphonyl piperazine, methylsulfonyl piperazine, piperazine, 1-methylsulfonyl, 1-n-methanesulfonylpiperazine, 4-methylsulfonylpiperazine, n-mesylpiperazine
PubChem CID 709161
IUPAC Name 4-methanesulfonylpiperazin-1-ium
SMILES CS(=O)(=O)N1CC[NH2+]CC1

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 164.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Methylsulfonyl)piperazine