missing translation for 'onlineSavingsMsg'
Learn More

1-(Methylsulfonyl)piperazine, 97%

Catalog No. AAH6387022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6387022 100 g
AAH6387006 5 g
AAH6387014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6387022 Supplier Thermo Scientific Chemicals Supplier No. H6387022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55276-43-2 | C5H13N2O2S | 165.23 g/mol

1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 55276-43-2
Molecular Formula C5H13N2O2S
Molecular Weight (g/mol) 165.23
MDL Number MFCD02153225
InChI Key ZZAKLGGGMWORRT-UHFFFAOYSA-O
Synonym 1-methylsulfonyl piperazine, 1-methanesulfonylpiperazine, 1-methanesulfonyl-piperazine, 1-methylsulfonyl-piperazine, 1-methylsulphonyl piperazine, methylsulfonyl piperazine, piperazine, 1-methylsulfonyl, 1-n-methanesulfonylpiperazine, 4-methylsulfonylpiperazine, n-mesylpiperazine
PubChem CID 709161
IUPAC Name 1-methylsulfonylpiperazine
SMILES CS(=O)(=O)N1CC[NH2+]CC1

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 164.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Methylsulfonyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only