Learn More
1-Methylsulfonyl-1H-benzotriazole, 97%
CAS: 37073-15-7 | C7H7N3O2S | 197.21 g/mol
$63.71 - $739.66
Chemical Identifiers
| CAS | 37073-15-7 |
|---|---|
| Molecular Formula | C7H7N3O2S |
| Molecular Weight (g/mol) | 197.21 |
| MDL Number | MFCD00595638 |
| InChI Key | OMMZSDGGKVRGGP-UHFFFAOYSA-N |
| Synonym | 1-methylsulfonyl-1h-benzotriazole, 1-methylsulfonyl-1h-benzo d 1,2,3 triazole, 1-methanesulfonyl-1h-benzotriazole, 1-methanesulfonyl-1,2,3-benzotriazole, acmc-20ap9j, cambridge id 5284919, methylsulfonyl benzotriazole, 1-methylsulfonyl benzotriazole, 1-methylsulphonyl-1h-benzotriazole |
| PubChem CID | 792044 |
| SMILES | CS(=O)(=O)N1N=NC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5338603
|
Thermo Scientific Chemicals
H5338603 |
1 g |
Each for $63.71
|
|
|||||
|
AAH5338606
|
Thermo Scientific Chemicals
H5338606 |
5 g |
Each for $218.90
|
|
|||||
|
AAH5338614
|
Thermo Scientific Chemicals
H5338614 |
25 g |
Each for $739.66
|
|
|||||
Description
1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
| 37073-15-7 | |
| 197.21 | |
| OMMZSDGGKVRGGP-UHFFFAOYSA-N | |
| 792044 |
| C7H7N3O2S | |
| MFCD00595638 | |
| 1-methylsulfonyl-1h-benzotriazole, 1-methylsulfonyl-1h-benzo d 1,2,3 triazole, 1-methanesulfonyl-1h-benzotriazole, 1-methanesulfonyl-1,2,3-benzotriazole, acmc-20ap9j, cambridge id 5284919, methylsulfonyl benzotriazole, 1-methylsulfonyl benzotriazole, 1-methylsulphonyl-1h-benzotriazole | |
| CS(=O)(=O)N1N=NC2=CC=CC=C12 |
Specifications
| 37073-15-7 | |
| C7H7N3O2S | |
| 1 g | |
| OMMZSDGGKVRGGP-UHFFFAOYSA-N | |
| 1-methanesulfonyl-1H-1,2,3-benzotriazole | |
| 792044 | |
| 97% |
| 106°C to 108°C (decomposition) | |
| MFCD00595638 | |
| 1-methylsulfonyl-1h-benzotriazole, 1-methylsulfonyl-1h-benzo d 1,2,3 triazole, 1-methanesulfonyl-1h-benzotriazole, 1-methanesulfonyl-1,2,3-benzotriazole, acmc-20ap9j, cambridge id 5284919, methylsulfonyl benzotriazole, 1-methylsulfonyl benzotriazole, 1-methylsulphonyl-1h-benzotriazole | |
| CS(=O)(=O)N1N=NC2=CC=CC=C12 | |
| 197.21 | |
| 197.21 | |
| 1-Methylsulfonyl-1H-benzotriazole |
Safety and Handling
GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only