missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrolidine 98.0+%, TCI America™

Catalog No. M041525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M041525ML 25 mL
M0415100ML 100 mL
M0415500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M041525ML Supplier TCI America Supplier No. M041525ML

Chemical Identifiers

CAS 120-94-5
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00003173
InChI Key AVFZOVWCLRSYKC-UHFFFAOYSA-N
Synonym n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine
PubChem CID 8454
IUPAC Name 1-methylpyrrolidine
SMILES CN1CCCC1

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 mL
UN Number 2924
Formula Weight 85.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrolidine
EINECSNumber (5)-6168
RTECSNumber UY1420500
TSCA Yes