missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrolidine, 98%

Catalog No. p-7027218
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2379922 100 g
AAB2379936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2379922 Supplier Thermo Scientific Chemicals Supplier No. B2379922

CAS: 120-94-5 | C5H11N | 85.15 g/mol

1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 120-94-5
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00003173
InChI Key AVFZOVWCLRSYKC-UHFFFAOYSA-N
Synonym n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine
PubChem CID 8454
IUPAC Name 1-methylpyrrolidine
SMILES CN1CCCC1

Specifications

Melting Point °C to 90°C
Density 0.819
Boiling Point 80°C to 81°C
Flash Point -18°C (0°F)
Assay Percent Range 0.98
Refractive Index 1.4247
Quantity 100 g
UN Number UN3286
Solubility Information Miscible with water.
Formula Weight 85.15
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrolidine
Show More Show Less
Hazard Category H225-H301-H314-H332-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302-H332
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 204-438-5
RTECSNumber UY1420500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only