missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrolidine, 98%

Catalog No. p-4509816
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127611000 100 g Glass bottle
AC127615000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC127611000 Supplier Thermo Scientific Chemicals Supplier No. 127611000

CAS: 120-94-5 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-94-5
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00003173
InChI Key AVFZOVWCLRSYKC-UHFFFAOYSA-N
Synonym n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine
PubChem CID 8454
IUPAC Name 1-methylpyrrolidine
SMILES CN1CCCC1

Specifications

Melting Point -90.0°C
Color Colorless
Density 0.8190g/mL
Boiling Point 80.0°C to 81.0°C
Flash Point −21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4237 to 1.4257
Quantity 100 g
Beilstein 20,4
Specific Gravity 0.819
Solubility Information Solubility in water: fully miscible.
Viscosity 0.48 mPa.s (20°C)
Formula Weight 85.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 204-438-5

RUO – Research Use Only