missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole 99.0+%, TCI America™

Catalog No. p-7132154
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M041425ML 25 mL
M0414500ML 500 mL
2 options

Catalog No. M041425ML

Supplier: TCI America M041425ML

Only null left

Chemical Identifiers

CAS 96-54-8
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00005345
InChI Key OXHNLMTVIGZXSG-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole, n-methylpyrrole, n-methyl pyrrole, 1h-pyrrole, 1-methyl, pyrrole, 1-methyl, methylpyrrole, n-methylpyrrol, 1-methylpyrrol, unii-ng5gpn98zg, ccris 2934
PubChem CID 7304
IUPAC Name 1-methylpyrrole
SMILES CN1C=CC=C1

Specifications

Melting Point -57°C
Color Yellow
Boiling Point 113°C
Quantity 25 mL
UN Number 1993
Formula Weight 81.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrole
EINECSNumber (5)-3371
RTECSNumber UX9640000
TSCA Yes