missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole 99.0+%, TCI America™

Catalog No. M0414500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0414500ML 500 mL
M041425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0414500ML Supplier TCI America Supplier No. M0414500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-54-8
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00005345
InChI Key OXHNLMTVIGZXSG-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole, n-methylpyrrole, n-methyl pyrrole, 1h-pyrrole, 1-methyl, pyrrole, 1-methyl, methylpyrrole, n-methylpyrrol, 1-methylpyrrol, unii-ng5gpn98zg, ccris 2934
PubChem CID 7304
IUPAC Name 1-methylpyrrole
SMILES CN1C=CC=C1

Specifications

Melting Point -57°C
Color Yellow
Boiling Point 113°C
Quantity 500 mL
UN Number 1993
Formula Weight 81.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrole
EINECSNumber (5)-3371
RTECSNumber UX9640000
TSCA Yes