missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole, 99%

Catalog No. p-7025782
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21451AE 100 mL
AAB21451AP 500 mL
AAB214510F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB21451AE Supplier Thermo Scientific Chemicals Supplier No. B21451AE
Only null left

CAS: 96-54-8 | C5H7N | 81.118 g/mol

1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents such as alcohol and so on, but insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Stable under normal temperatures and pressures.

Chemical Identifiers

CAS 96-54-8
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00005345
InChI Key OXHNLMTVIGZXSG-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole, n-methylpyrrole, n-methyl pyrrole, 1h-pyrrole, 1-methyl, pyrrole, 1-methyl, methylpyrrole, n-methylpyrrol, 1-methylpyrrol, unii-ng5gpn98zg, ccris 2934
PubChem CID 7304
IUPAC Name 1-methylpyrrole
SMILES CN1C=CC=C1

Specifications

Melting Point -57°C
Density 0.912
Boiling Point 112°C to 113°C
Flash Point 15°C (59°F)
Refractive Index 1.488
Quantity 100 mL
UN Number UN1993
Beilstein 104181
Solubility Information Soluble in organic solvents such as alcohol and so on,but insoluble in water.
Formula Weight 81.12
Percent Purity 99%
Chemical Name or Material 1-Methylpyrrole
Show More Show Less
Hazard Category H225-H302-H315-H319-H335
Hazard Statement GHS H Statement
H225-H301-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 202-513-7
RTECSNumber UX9640000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only