missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpyrrole, 99%
Description
1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents such as alcohol and so on, but insoluble in water.
Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Stable under normal temperatures and pressures.
1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents such as alcohol and so on, but insoluble in water.
Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Stable under normal temperatures and pressures.
Specifications
Specifications
| Melting Point | -57°C |
| Density | 0.912 |
| Boiling Point | 112°C to 113°C |
| Flash Point | 15°C (59°F) |
| Refractive Index | 1.488 |
| Quantity | 500 mL |
| UN Number | UN1993 |
| Beilstein | 104181 |
| Solubility Information | Soluble in organic solvents such as alcohol and so on,but insoluble in water. |
| Formula Weight | 81.12 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H315-H319-H335 |
| Hazard Statement | GHS H StatementH225-H301-H315-H319-H335Highly flammable liquid and vapour.Toxic if swallowed.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 202-513-7 |
| RTECSNumber | UX9640000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only