missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrazole-5-carboxaldehyde 95.0+%, TCI America™

Catalog No. M21791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M21791G Supplier TCI America Supplier No. M21791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27258-33-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD03419801
InChI Key RAJRANFZSWDUJZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-carbaldehyde, 2-methyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-1h-pyrazole, 1-methylpyrazole-5-carboxaldehyde, 5-formyl-1-methylpyrazole, 1-methylpyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-methyl, 1-methyl-1h-pyrazole-5-carboxaldehyde, pubchem16289, acmc-209gw7
PubChem CID 7019414
IUPAC Name 1-methyl-1H-pyrazole-5-carbaldehyde
SMILES CN1N=CC=C1C=O

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrazole-5-carboxaldehyde