missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. M22241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22241G 1 g
M22245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22241G Supplier TCI America Supplier No. M22241G

Chemical Identifiers

CAS 5952-92-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00159641
InChI Key UPPPWUOZCSMDTR-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-methyl, 4-carboxy-1-methyl-1h-pyrazole, 1-methyl-pyrazole-4-carboxylic acid, 1-methylpyrazole-4-carboxylicacid, zlchem 1243, pubchem16306, pubchem20860, acmc-209me3
PubChem CID 643160
ChEBI CHEBI:74741
IUPAC Name 1-methylpyrazole-4-carboxylic acid
SMILES CN1C=C(C=N1)C(=O)O

Specifications

Melting Point 207°C
Color White
Quantity 1 g
Formula Weight 126.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrazole-4-carboxylic Acid