missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine 99.0+%, TCI America™

Catalog No. M0403500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0403500ML 500 mL
M040325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0403500ML Supplier TCI America Supplier No. M0403500ML

Chemical Identifiers

CAS 626-67-5
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00006491
InChI Key QDUXDCXILAPLAG-UHFFFAOYSA-N
Synonym n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
PubChem CID 12291
IUPAC Name hydrogen 1-methylpiperidine chloride
SMILES [H+].[Cl-].CN1CCCCC1

Specifications

Color Yellow
Boiling Point 107°C
Quantity 500 mL
UN Number 2399
Formula Weight 99.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpiperidine
EINECSNumber (5)-0767
RTECSNumber TN1225000
TSCA Yes