missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine 98.0+%, TCI America™

Catalog No. p-7132233
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
M055325ML 25 mL
M0553100ML 100 mL
M0553500ML 500 mL
3 options

Catalog No. M055325ML

Supplier: TCI America M055325ML

Only null left

Chemical Identifiers

CAS 109-01-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005966
InChI Key PVOAHINGSUIXLS-UHFFFAOYSA-N
Synonym n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
PubChem CID 53167
IUPAC Name 1-methylpiperazine
SMILES CN1CCNCC1

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 137°C
Quantity 25 mL
UN Number 2924
Formula Weight 100.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpiperazine
EINECSNumber (5)-0954
RTECSNumber TM1225000
TSCA Yes