missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine 98.0+%, TCI America™

Catalog No. M055325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M055325ML 25 mL
M0553100ML 100 mL
M0553500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M055325ML Supplier TCI America Supplier No. M055325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-01-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005966
InChI Key PVOAHINGSUIXLS-UHFFFAOYSA-N
Synonym n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
PubChem CID 53167
IUPAC Name 1-methylpiperazine
SMILES CN1CCNCC1

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 137°C
Quantity 25 mL
UN Number 2924
Formula Weight 100.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpiperazine
EINECSNumber (5)-0954
RTECSNumber TM1225000
TSCA Yes