missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022863
Change view
Click to view available options
Quantity:
15 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022863 15 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022863 Supplier Merck Emd Millipore Supplier No. BP1118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 109-01-3
Density 0.903 g/mL (at 25°C (literature))
Boiling Point 138°C (literature)
Flash Point 39°C (Closed Cup)
Molecular Formula C5H12N2
Refractive Index n20/D 1.466 (literature)
Linear Formula C5H12N2
MDL Number MFCD00005966
Quantity 15 mg
Vapor Density 3.5 (vs air)
Formula Weight 100.16
Grade BP
Show More Show Less