missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, 99%

CAS: 109-01-3 | C5H12N2 | 100.16 g/mol

Supplier:  Thermo Scientific Chemicals 156911000

 View more versions of this product

Catalog No. AC156911000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methylpiperazine
-6.0°C
138.0°C
Authentic
Glass bottle
1.4645 to 1.4665
0.903
Solubility in water: soluble. Other solubilities: soluble in ethanol and aceton
CN1CCNCC1
100.16
4 mPa.s (20°C)
99%
109-01-3
0.9030g/mL
42°C
98.5% min. (GC)
C5H12N2
100 g
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
PVOAHINGSUIXLS-UHFFFAOYSA-N
1-methylpiperazine
53167
100.16
Liquid

Chemical Identifiers

109-01-3
100.16
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
1-methylpiperazine
C5H12N2
PVOAHINGSUIXLS-UHFFFAOYSA-N
53167
CN1CCNCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 203-639-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only