missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, 99%

Catalog No. AC156915000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156915000 500 g Glass bottle
AC156910050 5 g Glass bottle
AC156911000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156915000 Supplier Thermo Scientific Chemicals Supplier No. 156915000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-01-3 | C5H12N2 | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-01-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
InChI Key PVOAHINGSUIXLS-UHFFFAOYSA-N
Synonym n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
PubChem CID 53167
IUPAC Name 1-methylpiperazine
SMILES CN1CCNCC1

Specifications

Melting Point -6.0°C
Density 0.9030g/mL
Boiling Point 138.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4645 to 1.4665
Quantity 500 g
Specific Gravity 0.903
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and aceton
Viscosity 4 mPa.s (20°C)
Formula Weight 100.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Methylpiperazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 203-639-5

RUO – Research Use Only