missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole 96.0+%, TCI America™

Supplier:  TCI America M056125G

 View more versions of this product

Catalog No. M056125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Methylindole
246°C
MFCD00005800
1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
CN1C=CC2=CC=CC=C21
131.178
131.18
Liquid
603-76-9
C9H9N
25 g
BLRHMMGNCXNXJL-UHFFFAOYSA-N
1-methylindole
11781
≥96.0% (GC)

Chemical Identifiers

603-76-9
131.178
BLRHMMGNCXNXJL-UHFFFAOYSA-N
11781
CN1C=CC2=CC=CC=C21
C9H9N
MFCD00005800
1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
1-methylindole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions