missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole 96.0+%, TCI America™

Catalog No. M05615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05615G 5 g
M056125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. M05615G missing translation for 'mfr' TCI America missing translation for 'supplierNo' M05615G

Chemical Identifiers

CAS 603-76-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005800
InChI Key BLRHMMGNCXNXJL-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
PubChem CID 11781
IUPAC Name 1-methylindole
SMILES CN1C=CC2=CC=CC=C21

Specifications

Boiling Point 246°C
Quantity 5 g
Formula Weight 131.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylindole