missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole, 98%

Catalog No. p-7020594
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1260509 10 g
AAA1260518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1260509 Supplier Thermo Scientific Chemicals Supplier No. A1260509
Only null left

CAS: 603-76-9 | C9H9N | 131.178 g/mol

1-Methylindole is useful for the determination of association constant for the electron-donor-acceptor complexes with 1-(2,4,6-trinitrophenyl) propan-2-one. It acts as a reactant for the preparation of polycyclic derivatives of indoles, bisindole derivatives and pharmaceutically active 2-oxo-1-pyrrolidine analogues. It is also used as a reactant for the preparation of Positron Emission Tomography (PET) imaging agents of protein kinase C (PKC) and glycogen synthase kinase-3 (GSK-3).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindole is useful for the determination of association constant for the electron-donor-acceptor complexes with 1-(2,4,6-trinitrophenyl) propan-2-one. It acts as a reactant for the preparation of polycyclic derivatives of indoles, bisindole derivatives and pharmaceutically active 2-oxo-1-pyrrolidine analogues. It is also used as a reactant for the preparation of Positron Emission Tomography (PET) imaging agents of protein kinase C (PKC) and glycogen synthase kinase-3 (GSK-3).

Solubility
Immiscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 603-76-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005800
InChI Key BLRHMMGNCXNXJL-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
PubChem CID 11781
IUPAC Name 1-methylindole
SMILES CN1C=CC2=CC=CC=C21

Specifications

Density 1.048
Boiling Point 237°C to 239°C
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.6065
Quantity 10 g
Beilstein 111026
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 1-Methylindole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 210-057-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only