missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-carboxaldehyde, 98+%

Catalog No. AAL0221203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0221203 1 g
AAL0221206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0221203 Supplier Thermo Scientific Chemicals Supplier No. L0221203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19012-03-4 | C10H9NO | 159.19 g/mol

1-Methylindole-3-carboxaldehyde is used as a reactant for preparation of nitroolefins and β-nitroalcohols via microwave- or ultrasound-assisted Henry reactions, reactant for synthesis of quinolinones via three-component Ugi reaction, reactant for synthesis of α-ketoamides as inhibitors of Dengue virus protease with antiviral activity in cell-culture, reactant for preparation of thiazolopyrimidinones as inhibitors of Bcl-2 proteins, reactant for preparation of vinylindoles via Peterson olefination or olefination with Nysted reagent, reactant for preparation of indolyl alkenes from microwave-enhanced Knoevenagel condensation as antibacterial agents 1-Methylindole-3-carboxaldehyde may be used in the synthesis of (Z)-3-(1-methyl-1H-indol-3-yl)-2-(thiophen-3-yl)acrylonitrile, via base-catalyzed condensation with thiophene-3-acetonitrile. It was also used in the preparation of monomer, required for the synthesis of poly(3-vinyl-1-methylindole).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindole-3-carboxaldehyde is used as a reactant for preparation of nitroolefins and β-nitroalcohols via microwave- or ultrasound-assisted Henry reactions, reactant for synthesis of quinolinones via three-component Ugi reaction, reactant for synthesis of α-ketoamides as inhibitors of Dengue virus protease with antiviral activity in cell-culture, reactant for preparation of thiazolopyrimidinones as inhibitors of Bcl-2 proteins, reactant for preparation of vinylindoles via Peterson olefination or olefination with Nysted reagent, reactant for preparation of indolyl alkenes from microwave-enhanced Knoevenagel condensation as antibacterial agents 1-Methylindole-3-carboxaldehyde may be used in the synthesis of (Z)-3-(1-methyl-1H-indol-3-yl)-2-(thiophen-3-yl)acrylonitrile, via base-catalyzed condensation with thiophene-3-acetonitrile. It was also used in the preparation of monomer, required for the synthesis of poly(3-vinyl-1-methylindole).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.

Chemical Identifiers

CAS 19012-03-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00014570
InChI Key KXYBYRKRRGSZCX-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-3-carbaldehyde, 1-methylindole-3-carboxaldehyde, 1-methyl-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methylindole, 1-methyl-3-formylindole, n-methyl-3-formylindole, indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methyl-1h-indole, n-methyl-3-indolecarboxaldehyde
PubChem CID 87894
IUPAC Name 1-methylindole-3-carbaldehyde
SMILES CN1C=C(C=O)C2=CC=CC=C12

Specifications

Melting Point 67°C to 71°C
Quantity 1 g
Beilstein 121302
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 159.19
Percent Purity ≥98%
Chemical Name or Material 1-Methylindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-750-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only