missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylindole-2-carboxylic Acid 98.0+%, TCI America™
$91.33 - $262.83
Chemical Identifiers
| CAS | 16136-58-6 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.187 |
| MDL Number | MFCD00005801 |
| InChI Key | MAHAMBLNIDMREX-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa |
| PubChem CID | 27695 |
| IUPAC Name | 1-methylindole-2-carboxylic acid |
| SMILES | CN1C2=CC=CC=C2C=C1C(=O)O |
Chemical Identifiers
| 16136-58-6 | |
| 175.187 | |
| MAHAMBLNIDMREX-UHFFFAOYSA-N | |
| 27695 | |
| CN1C2=CC=CC=C2C=C1C(=O)O |
| C10H9NO2 | |
| MFCD00005801 | |
| 1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa | |
| 1-methylindole-2-carboxylic acid |
Specifications
| 16136-58-6 | |
| C10H9NO2 | |
| 1 g | |
| MAHAMBLNIDMREX-UHFFFAOYSA-N | |
| 1-methylindole-2-carboxylic acid | |
| 27695 | |
| ≥98.0% (GC,T) | |
| 1-Methylindole-2-carboxylic Acid |
| 212°C | |
| MFCD00005801 | |
| 1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa | |
| CN1C2=CC=CC=C2C=C1C(=O)O | |
| 175.187 | |
| 175.19 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NL6012400
TSCA : No